Product Details

SNP ID
hCV15930365
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:113930584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACTTTCAAAGCTTTCGTTAAAAAA[A/C]CAAAAGGGAGGGGGAAGGTGGAGAA
Phenotype
MIM: 608003
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
HIPK1 PubMed Links

Gene Details

Gene
HIPK1
Gene Name
homeodomain interacting protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152696.3 Intron NP_689909.2
NM_181358.2 Intron NP_852003.1
NM_198268.2 Intron NP_938009.1
NM_198269.2 Intron NP_938010.1
Gene
HIPK1-AS1
Gene Name
HIPK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details