Product Details

SNP ID
rs2783637
Assay Type
Validated
NCBI dbSNP Submissions
45
Location
Chr.1:26112735 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
TTAAATCTATCCTGTAGACTTAAAG[C/T]AGTGGATCCCAACTTCTCAGGCTCT
Phenotype
MIM: 610785
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDIK1L PubMed Links

Gene Details

Gene
PDIK1L
Gene Name
PDLIM1 interacting kinase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243532.1 Intron NP_001230461.1
NM_001243533.1 Intron NP_001230462.1
NM_152835.4 Intron NP_690048.1
XM_005245743.4 Intron XP_005245800.1
XM_006710381.3 Intron XP_006710444.1
XM_011540792.2 Intron XP_011539094.1

View Full Product Details