Product Details

SNP ID
rs2783708
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:26159523 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCCTGGGACCTTGGAATCTGAC[C/G]CACTGCCAAGGAGACTCCCTGGGAG
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM110D PubMed Links

Gene Details

Gene
FAM110D
Gene Name
family with sequence similarity 110 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024869.2 Intron NP_079145.2

View Full Product Details