Product Details

SNP ID
rs2211194
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:243499203 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTATCTGCGTAAAACTAAGAGAC[A/C]TCAGTACACCCAATCCCCTCTTCCA
Phenotype
MIM: 611223 MIM: 613524
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
AKT3 PubMed Links

Gene Details

Gene
AKT3
Gene Name
AKT serine/threonine kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206729.1 Intron NP_001193658.1
NM_005465.4 Intron NP_005456.1
NM_181690.2 Intron NP_859029.1
Gene
SDCCAG8
Gene Name
serologically defined colon cancer antigen 8
There are no transcripts associated with this gene.

View Full Product Details