Product Details

SNP ID
rs2232411
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64292811 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGATAGGAGGGGGACTGGGTGTGT[A/C]CAGCCAGAGGGATGTCTGTGGACAG
Phenotype
MIM: 603167 MIM: 605720
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
BAD PubMed Links

Gene Details

Gene
BAD
Gene Name
BCL2 associated agonist of cell death
There are no transcripts associated with this gene.

Gene
GPR137
Gene Name
G protein-coupled receptor 137
There are no transcripts associated with this gene.

Gene
KCNK4
Gene Name
potassium two pore domain channel subfamily K member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317090.1 Intron NP_001304019.1
NM_033310.2 Intron NP_201567.1
Gene
KCNK4-TEX40
Gene Name
KCNK4-TEX40 readthrough
There are no transcripts associated with this gene.

Gene
TEX40
Gene Name
testis expressed 40
There are no transcripts associated with this gene.

View Full Product Details