Product Details
- SNP ID
-
rs2232568
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:48568698 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCCAGTGGGTTCTCTAAACTCAAGG[C/T]TTGTGCCTGTTGTCTTTCTATATGA
- Phenotype
-
MIM: 149750
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LALBA
PubMed Links
Gene Details
- Gene
- LALBA
- Gene Name
- lactalbumin alpha
View Full Product Details