Product Details

SNP ID
rs2232672
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:27621584 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGCCACTGGGAAGGGAAGTGATT[G/T]GGGGGCGGCAGGTAAGGCCGCGCCC
Phenotype
MIM: 609733 MIM: 613715
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LNX2 PubMed Links

Gene Details

Gene
LNX2
Gene Name
ligand of numb-protein X 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153371.3 646 Intron NP_699202.1
XM_011534995.2 646 Intron XP_011533297.1
XM_017020434.1 646 Intron XP_016875923.1
Gene
POLR1D
Gene Name
RNA polymerase I subunit D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206559.1 646 Intron NP_001193488.1
NM_015972.3 646 Intron NP_057056.1
NM_152705.2 646 Intron NP_689918.1
XM_005266412.1 646 UTR 5 XP_005266469.1
XM_005266414.2 646 UTR 5 XP_005266471.1
XM_017020622.1 646 Intron XP_016876111.1
XM_017020623.1 646 UTR 5 XP_016876112.1

View Full Product Details