Product Details
- SNP ID
-
rs2925651
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:72125641 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TAATAAAGTACTGATAGATATTCCT[C/T]CAAAAAAAAGAAAGACTATTTCGTG
- Phenotype
-
MIM: 604875
MIM: 608659
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
MYO9A
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs149649570] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- MYO9A
- Gene Name
- myosin IXA
There are no transcripts associated with this gene.
- Gene
- SENP8
- Gene Name
- SUMO/sentrin peptidase family member, NEDD8 specific
View Full Product Details