Product Details

SNP ID
rs2232766
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8308863 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGTGAGTAAATGAATAAATGAAAC[A/G]TCCATTCTCGGAGTCGGGTTGCTTG
Phenotype
MIM: 606475 MIM: 602139
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CD320 PubMed Links

Gene Details

Gene
CD320
Gene Name
CD320 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001165895.1 Intron NP_001159367.1
NM_016579.3 Intron NP_057663.1
Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005001.4 Intron NP_004992.2

View Full Product Details