Product Details

SNP ID
rs2232815
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:171783658 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CTTCTGAAAATGTGACTCAGCCAAG[A/G]TGAGACTTGATTCCTTGAAGTACCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
METTL13 PubMed Links

Gene Details

Gene
METTL13
Gene Name
methyltransferase like 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001007239.1 Intron NP_001007240.1
NM_014955.2 Intron NP_055770.1
NM_015935.4 Intron NP_057019.3
XM_017001455.1 Intron XP_016856944.1
XM_017001456.1 Intron XP_016856945.1
XM_017001457.1 Intron XP_016856946.1
XM_017001458.1 Intron XP_016856947.1
XM_017001459.1 Intron XP_016856948.1

View Full Product Details