Product Details

SNP ID
rs2232979
Assay Type
Functionally tested
NCBI dbSNP Submissions
13
Location
Chr.1:23792893 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGGCCAGCAGGCTCGGGGTAGTAA[A/C]CTGTTTGGCTGCTACCTGGGTTCCC
Phenotype
MIM: 606953 MIM: 613898 MIM: 616143
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GALE PubMed Links

Gene Details

Gene
GALE
Gene Name
UDP-galactose-4-epimerase
There are no transcripts associated with this gene.

Gene
HMGCL
Gene Name
3-hydroxymethyl-3-methylglutaryl-CoA lyase
There are no transcripts associated with this gene.

Gene
LYPLA2
Gene Name
lysophospholipase II
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007260.2 Intron NP_009191.1
XM_005245728.4 Intron XP_005245785.1
XM_017000165.1 Intron XP_016855654.1
Gene
PITHD1
Gene Name
PITH domain containing 1
There are no transcripts associated with this gene.

View Full Product Details