Product Details

SNP ID
rs2233144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:16111928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGAAGACCTACGATTACCTGTT[C/T]AAGCTGCTGCTGATCGGGGACTCGG
Phenotype
MIM: 165040 MIM: 600317
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB8A PubMed Links

Gene Details

Gene
RAB8A
Gene Name
RAB8A, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005370.4 37 Silent Mutation TTC,TTT F9F NP_005361.2
XM_011528020.2 37 Silent Mutation TTC,TTT F9F XP_011526322.1
Gene
TPM4
Gene Name
tropomyosin 4
There are no transcripts associated with this gene.

View Full Product Details