Product Details

SNP ID
rs2233197
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:6213701 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCACCCACCCCTCCGTAGCCTCCC[C/T]GCCTTGTCGTAGGTGCCCCCCCACC
Phenotype
MIM: 159556
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MLLT1 PubMed Links

Gene Details

Gene
MLLT1
Gene Name
MLLT1, super elongation complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005934.3 Intron NP_005925.2
XM_011528021.1 Intron XP_011526323.1
XM_011528022.1 Intron XP_011526324.1
XM_011528023.1 Intron XP_011526325.1
XM_017026819.1 Intron XP_016882308.1

View Full Product Details