Product Details

SNP ID
rs2233242
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:60384945 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACATGACTTTCTCTAGAAGCATATC[A/G]ATTGGCTCCTTGTAACATTTTTCCT
Phenotype
MIM: 606502
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MS4A7 PubMed Links

Gene Details

Gene
MS4A7
Gene Name
membrane spanning 4-domains A7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021201.4 Intron NP_067024.1
NM_206938.1 Intron NP_996821.1
NM_206939.1 Intron NP_996822.1
NM_206940.1 Intron NP_996823.1

View Full Product Details