Product Details

SNP ID
rs2233446
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:85799708 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCGGCCTTGCTCTCTTCCGGTCGC[G/T]GGACACCGGGTGTAGAGGGCGGTCG
Phenotype
MIM: 123864 MIM: 604886
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COX4I1 PubMed Links

Gene Details

Gene
COX4I1
Gene Name
cytochrome c oxidase subunit 4I1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318786.1 37 UTR 5 NP_001305715.1
NM_001318788.1 37 UTR 5 NP_001305717.1
NM_001318794.1 37 UTR 5 NP_001305723.1
NM_001318797.1 37 UTR 5 NP_001305726.1
NM_001318802.1 37 UTR 5 NP_001305731.1
NM_001861.4 37 UTR 5 NP_001852.1
Gene
EMC8
Gene Name
ER membrane protein complex subunit 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142288.1 37 Intron NP_001135760.1
NM_006067.4 37 Intron NP_006058.1
XM_011522813.2 37 UTR 5 XP_011521115.1
XM_017022867.1 37 Intron XP_016878356.1
Gene
LOC101928557
Gene Name
uncharacterized LOC101928557
There are no transcripts associated with this gene.

View Full Product Details