Product Details
- SNP ID
-
rs2233582
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:127611832 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACTTTGGGTTAGTACTTTCTTGAC[A/G]TGAACACTGTGGGGCCCTGGAGAGC
- Phenotype
-
MIM: 167413
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PAX4
PubMed Links
Gene Details
- Gene
- PAX4
- Gene Name
- paired box 4
View Full Product Details