Product Details

SNP ID
rs2233868
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:153563870 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCCAGCGCCTGCTCCAGAGAACTG[C/G]ACATCATGGATCTGTGGCTGCAGAG
Phenotype
MIM: 176993
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
S100A2 PubMed Links

Gene Details

Gene
S100A2
Gene Name
S100 calcium binding protein A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005978.3 343 Missense Mutation TCC,TGC S2C NP_005969.1

View Full Product Details