Product Details

SNP ID
rs2233937
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:110297519 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTCTGCAGACTACTTCAAGTAT[C/G]CAAATGGGGAAGATTGCACCTAAAA
Phenotype
MIM: 603519
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SMNDC1 PubMed Links

Gene Details

Gene
SMNDC1
Gene Name
survival motor neuron domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005871.3 Intron NP_005862.1
XM_005269382.3 Intron XP_005269439.1

View Full Product Details