Product Details
- SNP ID
-
rs2233995
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141778496 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AATCCTCAATGAGTAAAGATGCTTC[A/G]GTTATTCTATTTCTCTGCTATTATT
- Phenotype
-
MIM: 604869
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2233996] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- TAS2R4
- Gene Name
- taste 2 receptor member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016944.1 |
8 |
Missense Mutation |
CAG,CGG |
Q3R |
NP_058640.1 |
View Full Product Details