Product Details

SNP ID
rs2234027
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:42745569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCTTCCGCCTTCTTGGGCAGCG[A/G]GGAGTCCTGTATGCTTTCAACTGCC
Phenotype
MIM: 602971
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GLTSCR1L PubMed Links

Gene Details

Gene
GLTSCR1L
Gene Name
GLTSCR1 like
There are no transcripts associated with this gene.

Gene
TBCC
Gene Name
tubulin folding cofactor C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003192.2 578 Missense Mutation CCG,TCG P169S NP_003183.1

View Full Product Details