Product Details

SNP ID
rs2234038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106035039 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTTCAGGGTGATTCTTCTGCCTCTG[A/G]GTTTCTGCTGAACTCTGTCTCACCT
Phenotype
MIM: 314200
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SERPINA7 PubMed Links

Gene Details

Gene
SERPINA7
Gene Name
serpin family A member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000354.5 Intron NP_000345.2
XM_005262180.3 Intron XP_005262237.1
XM_006724683.2 Intron XP_006724746.1

View Full Product Details