Product Details

SNP ID
rs2234046
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:35120241 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGGGTGAGCCAAACAAAACACTGC[C/G]GGAGAAACTACTGGCCACCAGCAAG
Phenotype
MIM: 186982
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
TCP11 PubMed Links

Gene Details

Gene
TCP11
Gene Name
t-complex 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001093728.2 1347 Missense Mutation CGC,GGC R358G NP_001087197.1
NM_001261817.1 1347 Missense Mutation CGC,GGC R353G NP_001248746.1
NM_001261818.1 1347 Missense Mutation CGC,GGC R312G NP_001248747.1
NM_001261819.1 1347 Missense Mutation CGC,GGC R307G NP_001248748.1
NM_001261820.1 1347 Missense Mutation CGC,GGC R282G NP_001248749.1
NM_001261821.1 1347 Missense Mutation CGC,GGC R282G NP_001248750.1
NM_018679.5 1347 Missense Mutation CGC,GGC R283G NP_061149.1
XM_005249339.2 1347 Missense Mutation CGC,GGC R204G XP_005249396.1
XM_011514830.1 1347 Missense Mutation CGC,GGC R461G XP_011513132.1
XM_011514831.1 1347 Missense Mutation CGC,GGC R204G XP_011513133.1
XM_011514832.1 1347 Missense Mutation CGC,GGC R204G XP_011513134.1

View Full Product Details