Product Details

SNP ID
rs2927457
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:44753690 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGAGGGCGGGGGACGCTAAGTCTGG[A/G]TCCCCGCAGAAGCCAAAAGGTTGGG
Phenotype
MIM: 109560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BCL3 PubMed Links

Gene Details

Gene
BCL3
Gene Name
B-cell CLL/lymphoma 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005178.4 Intron NP_005169.2
XM_011527197.2 Intron XP_011525499.2
XM_011527198.2 Intron XP_011525500.2
XM_011527200.2 Intron XP_011525502.1
XM_017027109.1 Intron XP_016882598.1
XM_017027110.1 Intron XP_016882599.1
Gene
MIR8085
Gene Name
microRNA 8085
There are no transcripts associated with this gene.

View Full Product Details