Product Details

SNP ID
rs2234269
Assay Type
Functionally tested
NCBI dbSNP Submissions
7
Location
Chr.1:46184435 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCTCCACAGGCTGATGACCAAGA[G/T]TCCCCTCGCCCTTGACACCAGGCCC
Phenotype
MIM: 606076 MIM: 606822 MIM: 613170
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PIK3R3 PubMed Links

Gene Details

Gene
PIK3R3
Gene Name
phosphoinositide-3-kinase regulatory subunit 3
There are no transcripts associated with this gene.

Gene
POMGNT1
Gene Name
protein O-linked mannose N-acetylglucosaminyltransferase 1 (beta 1,2-)
There are no transcripts associated with this gene.

Gene
TSPAN1
Gene Name
tetraspanin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005727.3 Intron NP_005718.2
XM_011540460.2 Intron XP_011538762.1

View Full Product Details