Product Details
- SNP ID
-
hCV15948679
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:102802596 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAAGGGGTAGTCTGTAGGAAAAT[A/G]AATTGTCAAAAAGAATACACCGCCT
- Phenotype
-
MIM: 602963
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LOC102723704
PubMed Links
Gene Details
- Gene
- LOC102723704
- Gene Name
- uncharacterized LOC102723704
There are no transcripts associated with this gene.
- Gene
- UBE2D3
- Gene Name
- ubiquitin conjugating enzyme E2 D3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001300795.1 |
813 |
Missense Mutation |
CAT,TAT |
H26Y |
NP_001287724.1 |
NM_003340.6 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_003331.1 |
NM_181886.3 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871615.1 |
NM_181887.2 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871616.1 |
NM_181888.3 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871617.1 |
NM_181889.2 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871618.1 |
NM_181890.2 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871619.1 |
NM_181891.2 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871620.1 |
NM_181892.3 |
813 |
Missense Mutation |
CAT,TAT |
H55Y |
NP_871621.1 |
NM_181893.2 |
813 |
Missense Mutation |
CAT,TAT |
H57Y |
NP_871622.1 |
View Full Product Details