Product Details
- SNP ID
-
rs2234417
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.13:29771996 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAGGCATACCAGAAATAGGCAGAA[C/T]TCATAGTTTGAATTTATCATTTTTT
- Phenotype
-
MIM: 604711
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
UBL3
PubMed Links
Gene Details
- Gene
- UBL3
- Gene Name
- ubiquitin like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_007106.3 |
|
Intron |
|
|
NP_009037.1 |
View Full Product Details