Product Details
- SNP ID
-
rs2234429
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:23562741 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATTGGCACTAAAGGTATGTTCTT[C/T]CTCGCCTCCTTGCTGGCCCACTGCC
- Phenotype
-
MIM: 612799
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
EARS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs78924422] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- EARS2
- Gene Name
- glutamyl-tRNA synthetase 2, mitochondrial
There are no transcripts associated with this gene.
- Gene
- UBFD1
- Gene Name
- ubiquitin family domain containing 1
View Full Product Details