Product Details

SNP ID
rs2234574
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1983941 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCGGGGGTGTCTGAGTGGTAGGGGC[A/G]ACAGCCGGAGGTGTGGTGGGTGTCA
Phenotype
MIM: 606026 MIM: 602952
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR943 PubMed Links

Gene Details

Gene
MIR943
Gene Name
microRNA 943
There are no transcripts associated with this gene.

Gene
NELFA
Gene Name
negative elongation factor complex member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005663.4 1485 Silent Mutation GTC,GTT V414V NP_005654.3
XM_017008589.1 1485 Silent Mutation GTC,GTT V442V XP_016864078.1
Gene
SCARNA22
Gene Name
small Cajal body-specific RNA 22
There are no transcripts associated with this gene.

Gene
WHSC1
Gene Name
Wolf-Hirschhorn syndrome candidate 1
There are no transcripts associated with this gene.

View Full Product Details