Product Details

SNP ID
rs2911153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81007741 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAACGTTACTTAAAATGGATCTTTT[C/T]GATCATGCGTTCCACAGATCTTGAG
Phenotype
MIM: 611509
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CENPN PubMed Links

Gene Details

Gene
CENPN
Gene Name
centromere protein N
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100624.2 Intron NP_001094094.2
NM_001100625.2 Intron NP_001094095.2
NM_001270473.1 Intron NP_001257402.1
NM_001270474.1 Intron NP_001257403.1
NM_018455.5 Intron NP_060925.2
XM_006721236.3 Intron XP_006721299.1
XM_017023456.1 Intron XP_016878945.1
Gene
CMC2
Gene Name
C-X9-C motif containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020188.3 Intron NP_064573.1
XM_017023461.1 Intron XP_016878950.1
XM_017023462.1 Intron XP_016878951.1
XM_017023463.1 Intron XP_016878952.1
XM_017023464.1 Intron XP_016878953.1
XM_017023465.1 Intron XP_016878954.1
XM_017023466.1 Intron XP_016878955.1
XM_017023467.1 Intron XP_016878956.1
XM_017023468.1 Intron XP_016878957.1
XM_017023469.1 Intron XP_016878958.1
XM_017023470.1 Intron XP_016878959.1

View Full Product Details