Product Details

SNP ID
rs2911160
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:81059552 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTTCATCAGAACATGTGTCTACA[A/T]AGCAACATCACTTGGCATTTAAAAT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C16orf46 PubMed Links

Gene Details

Gene
C16orf46
Gene Name
chromosome 16 open reading frame 46
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100873.1 Intron NP_001094343.1
NM_152337.2 Intron NP_689550.2
XM_005255792.2 Intron XP_005255849.1
XM_011522848.2 Intron XP_011521150.2
XM_011522849.2 Intron XP_011521151.2
XM_017022916.1 Intron XP_016878405.1
XM_017022917.1 Intron XP_016878406.1

View Full Product Details