Product Details

SNP ID
rs2073656
Assay Type
Validated
NCBI dbSNP Submissions
35
Location
Chr.1:161041565 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
GTCCAGGTCTTAATGATCACTAAGA[C/G]TCATGAGTGAAGGCTCACTGCCTAC
Phenotype
MIM: 614264 MIM: 616041 MIM: 191523
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ARHGAP30 PubMed Links

Gene Details

Gene
ARHGAP30
Gene Name
Rho GTPase activating protein 30
There are no transcripts associated with this gene.

Gene
TSTD1
Gene Name
thiosulfate sulfurtransferase like domain containing 1
There are no transcripts associated with this gene.

Gene
USF1
Gene Name
upstream transcription factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001276373.1 Intron NP_001263302.1
NM_007122.4 Intron NP_009053.1
NM_207005.2 Intron NP_996888.1

View Full Product Details