Product Details

SNP ID
rs2073440
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:50242317 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTGGAGTCCACATTGAGAGCTGCA[G/T]TCAGGAAAGCTGGGGACGCTGTAGA
Phenotype
MIM: 142704 MIM: 603247
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HDC PubMed Links

Gene Details

Gene
HDC
Gene Name
histidine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306146.1 2143 Missense Mutation GAA,GAC E611D NP_001293075.1
NM_002112.3 2143 Missense Mutation GAA,GAC E644D NP_002103.2
XM_017022094.1 2143 Missense Mutation GAA,GAC E679D XP_016877583.1
XM_017022095.1 2143 Missense Mutation GAA,GAC E646D XP_016877584.1
XM_017022096.1 2143 Missense Mutation GAA,GAC E603D XP_016877585.1
XM_017022097.1 2143 Missense Mutation GAA,GAC E600D XP_016877586.1
XM_017022098.1 2143 Missense Mutation GAA,GAC E535D XP_016877587.1
XM_017022099.1 2143 Intron XP_016877588.1
Gene
SLC27A2
Gene Name
solute carrier family 27 member 2
There are no transcripts associated with this gene.

View Full Product Details