Product Details

SNP ID
rs2235927
Assay Type
Functionally tested
NCBI dbSNP Submissions
72
Location
Chr.1:17068280 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
TATGGCTGACCAGGGGCATCTTTAC[A/G]CATTGAACTCTCAGGTCACAAGTAT
Phenotype
MIM: 607935
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PADI2 PubMed Links

Gene Details

Gene
PADI2
Gene Name
peptidyl arginine deiminase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_007365.2 2018 UTR 3 NP_031391.2
XM_017000148.1 2018 UTR 3 XP_016855637.1

View Full Product Details