Product Details

SNP ID
rs2236065
Assay Type
Validated
NCBI dbSNP Submissions
34
Location
Chr.1:231166701 on Build GRCh38
Set Membership
HapMap JSNP Validated
Context Sequence [VIC/FAM]
TTTTTCTCTGGTGTTACTGCAGCTT[A/G]CCTTCTGATTAGTGATTCTTAAGTA
Phenotype
MIM: 610584
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
TRIM67 PubMed Links
Additional Information
For this assay, SNP(s) [rs115461650,rs9431659] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
TRIM67
Gene Name
tripartite motif containing 67
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004342.3 Intron NP_001004342.3
NM_001300889.1 Intron NP_001287818.1
XM_011544192.2 Intron XP_011542494.1
XM_017001323.1 Intron XP_016856812.1

View Full Product Details