Product Details

SNP ID
rs2236078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:130833118 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAATCTCTCTGCACATCCTCCTTAA[C/T]AGACCCAATGAAATGAAATGACAGC
Phenotype
MIM: 603237
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPB41L2 PubMed Links

Gene Details

Gene
EPB41L2
Gene Name
erythrocyte membrane protein band 4.1 like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135554.1 Intron NP_001129026.1
NM_001135555.3 Intron NP_001129027.1
NM_001199388.2 Intron NP_001186317.1
NM_001252660.1 Intron NP_001239589.1
NM_001431.3 Intron NP_001422.1
XM_006715362.2 Intron XP_006715425.1
XM_011535524.1 Intron XP_011533826.1
XM_011535525.1 Intron XP_011533827.1
XM_011535527.1 Intron XP_011533829.1
XM_011535528.1 Intron XP_011533830.1
XM_011535529.1 Intron XP_011533831.1
XM_011535530.1 Intron XP_011533832.1
XM_011535534.1 Intron XP_011533836.1
XM_011535536.1 Intron XP_011533838.1
XM_017010349.1 Intron XP_016865838.1
XM_017010350.1 Intron XP_016865839.1
XM_017010351.1 Intron XP_016865840.1
XM_017010352.1 Intron XP_016865841.1
XM_017010353.1 Intron XP_016865842.1
XM_017010354.1 Intron XP_016865843.1
XM_017010355.1 Intron XP_016865844.1
XM_017010356.1 Intron XP_016865845.1
XM_017010357.1 Intron XP_016865846.1
XM_017010358.1 Intron XP_016865847.1
XM_017010359.1 Intron XP_016865848.1
XM_017010360.1 Intron XP_016865849.1
XM_017010361.1 Intron XP_016865850.1
XM_017010362.1 Intron XP_016865851.1
XM_017010363.1 Intron XP_016865852.1
XM_017010364.1 Intron XP_016865853.1
Gene
SMLR1
Gene Name
small leucine rich protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001195597.1 Intron NP_001182526.1

View Full Product Details