Product Details

SNP ID
rs2236213
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:52897719 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGGAGACCTGGGGGTACTGAAGG[C/T]CTGAAAAAGGCTGGGGTCAGAACAT
Phenotype
MIM: 605449
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GSTA3 PubMed Links

Gene Details

Gene
GSTA3
Gene Name
glutathione S-transferase alpha 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000847.4 Intron NP_000838.3
XM_006715072.3 Intron XP_006715135.1
XM_011514533.2 Intron XP_011512835.1
XM_017010794.1 Intron XP_016866283.1

View Full Product Details