Product Details

SNP ID
rs2236684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65571473 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAGCTCCCCCTGGGCTCTCGACC[G/T]GCGCCCCAGCCCCCAGTACCTCGTC
Phenotype
MIM: 616128 MIM: 606044
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EHBP1L1 PubMed Links

Gene Details

Gene
EHBP1L1
Gene Name
EH domain binding protein 1 like 1
There are no transcripts associated with this gene.

Gene
FAM89B
Gene Name
family with sequence similarity 89 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001098784.1 465 Intron NP_001092254.1
NM_001098785.1 465 Intron NP_001092255.1
NM_152832.2 465 Intron NP_690045.1
Gene
SSSCA1
Gene Name
Sjogren syndrome/scleroderma autoantigen 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303024.1 465 Silent Mutation CCG,CCT P76P NP_001289953.1
NM_006396.2 465 Silent Mutation CCG,CCT P113P NP_006387.1
Gene
SSSCA1-AS1
Gene Name
SSSCA1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details