Product Details

SNP ID
rs2236749
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:73166304 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGTTCTGCAGGCCCTGAGGGAGG[C/T]GGGGGGTGGGAAGTGAATGCGGGAA
Phenotype
MIM: 182452
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SSTR2 PubMed Links

Gene Details

Gene
SSTR2
Gene Name
somatostatin receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001050.2 Intron NP_001041.1

View Full Product Details