Product Details

SNP ID
rs2227252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218401718 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAATGGCGCCATCCCTAAGGTGCG[G/T]GGGGGCCAGGTGGGGCCACGGGGGC
Phenotype
MIM: 605323 MIM: 612152 MIM: 600266
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CTDSP1 PubMed Links

Gene Details

Gene
CTDSP1
Gene Name
CTD small phosphatase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206878.1 Intron NP_001193807.1
NM_021198.2 Intron NP_067021.1
NM_182642.2 Intron NP_872580.1
XM_011511569.2 Intron XP_011509871.1
XM_011511570.2 Intron XP_011509872.1
XM_017004615.1 Intron XP_016860104.1
XM_017004616.1 Intron XP_016860105.1
XM_017004617.1 Intron XP_016860106.1
XM_017004618.1 Intron XP_016860107.1
Gene
MIR26B
Gene Name
microRNA 26b
There are no transcripts associated with this gene.

Gene
SLC11A1
Gene Name
solute carrier family 11 member 1
There are no transcripts associated with this gene.

View Full Product Details