Product Details

SNP ID
rs2236942
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50211374 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTCCTAGACCTGGCCTTTACAGCT[C/G]TATCAGGTTTGTCTGTTGCCACAGA
Phenotype
MIM: 604437
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC38A3 PubMed Links

Gene Details

Gene
SLC38A3
Gene Name
solute carrier family 38 member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006841.5 Intron NP_006832.1
XM_006712954.2 Intron XP_006713017.1

View Full Product Details