Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001040177.2 | Intron | NP_001035267.1 | |||
NM_001271021.1 | Intron | NP_001257950.1 | |||
NM_001271025.1 | Intron | NP_001257954.1 | |||
XM_011519715.2 | Intron | XP_011518017.1 | |||
XM_011519718.2 | Intron | XP_011518020.1 | |||
XM_011519719.2 | Intron | XP_011518021.1 | |||
XM_011519720.2 | Intron | XP_011518022.1 | |||
XM_011519722.2 | Intron | XP_011518024.1 | |||
XM_011519724.2 | Intron | XP_011518026.1 | |||
XM_011519725.2 | Intron | XP_011518027.1 | |||
XM_011519729.2 | Intron | XP_011518031.1 | |||
XM_017016743.1 | Intron | XP_016872232.1 | |||
XM_017016744.1 | Intron | XP_016872233.1 | |||
XM_017016745.1 | Intron | XP_016872234.1 |