Product Details

SNP ID
rs2961665
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:178187093 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GCTTGTCCCCCTCCTCCTCATGCTC[C/G]TCCCTGTGCGAGTCCGGGTCACAGT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GMCL1P1 PubMed Links

Gene Details

Gene
GMCL1P1
Gene Name
germ cell-less, spermatogenesis associated 1 pseudogene 1
There are no transcripts associated with this gene.

View Full Product Details