Product Details

SNP ID
rs2270956
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:8695456 on Build GRCh38
Set Membership
Validated
Context Sequence [VIC/FAM]
ACATCTATCTCATCAGTAACAAGGC[C/T]ACTTACCCAAACCTCGGAGAAACTT
Phenotype
MIM: 603637 MIM: 611333 MIM: 140750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RPL27A PubMed Links

Gene Details

Gene
RPL27A
Gene Name
ribosomal protein L27a
There are no transcripts associated with this gene.

Gene
SNORA3B
Gene Name
small nucleolar RNA, H/ACA box 3B
There are no transcripts associated with this gene.

Gene
ST5
Gene Name
suppression of tumorigenicity 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005418.3 Intron NP_005409.3
NM_139157.2 Intron NP_631896.1
NM_213618.1 Intron NP_998783.1
XM_005253077.1 Intron XP_005253134.1
XM_005253083.2 Intron XP_005253140.1
XM_011520309.1 Intron XP_011518611.1
XM_011520310.1 Intron XP_011518612.1
XM_011520311.2 Intron XP_011518613.1
XM_011520312.1 Intron XP_011518614.1
XM_011520313.1 Intron XP_011518615.1
XM_011520314.1 Intron XP_011518616.1
XM_011520315.1 Intron XP_011518617.1
XM_011520316.1 Intron XP_011518618.1
XM_011520317.1 Intron XP_011518619.1
XM_011520318.1 Intron XP_011518620.1
XM_011520319.2 Intron XP_011518621.1
XM_011520320.1 Intron XP_011518622.1
XM_011520321.2 Intron XP_011518623.1
XM_011520322.1 Intron XP_011518624.1
XM_011520323.2 Intron XP_011518625.2
XM_011520324.2 Intron XP_011518626.2
XM_011520325.1 Intron XP_011518627.1
XM_011520326.1 Intron XP_011518628.1
XM_011520327.1 Intron XP_011518629.1
XM_011520328.1 Intron XP_011518630.1
XM_011520329.1 Intron XP_011518631.1
XM_017018182.1 Intron XP_016873671.1
XM_017018183.1 Intron XP_016873672.1
XM_017018184.1 Intron XP_016873673.1
XM_017018185.1 Intron XP_016873674.1
XM_017018186.1 Intron XP_016873675.1
XM_017018187.1 Intron XP_016873676.1

View Full Product Details