Product Details

SNP ID
rs2283678
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55012259 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAATATTCTTACAACATATTCAC[A/G]TGTACTTCAACATGCATCATAAAAA
Phenotype
MIM: 301300 MIM: 300773
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ALAS2 PubMed Links

Gene Details

Gene
ALAS2
Gene Name
5'-aminolevulinate synthase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000032.4 Intron NP_000023.2
NM_001037967.3 Intron NP_001033056.1
NM_001037968.3 Intron NP_001033057.1
XM_005261995.3 Intron XP_005262052.1
XM_011530771.1 Intron XP_011529073.1
XM_017029354.1 Intron XP_016884843.1
Gene
APEX2
Gene Name
apurinic/apyrimidinic endodeoxyribonuclease 2
There are no transcripts associated with this gene.

View Full Product Details