Product Details

SNP ID
rs2271587
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:3638873 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGGGGCCAAGAGCAGAGTCTTCCA[G/T]GGGGGCAGAGAGGAGGCCCCCTCAG
Phenotype
MIM: 601625 MIM: 610625
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ART1 PubMed Links

Gene Details

Gene
ART1
Gene Name
ADP-ribosyltransferase 1
There are no transcripts associated with this gene.

Gene
ART5
Gene Name
ADP-ribosyltransferase 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079536.1 1117 Intron NP_001073004.1
NM_001297668.1 1117 Silent Mutation NP_001284597.1
NM_053017.4 1117 Intron NP_443750.2
XM_005252779.3 1117 Silent Mutation XP_005252836.1
XM_006718136.3 1117 Silent Mutation XP_006718199.1
XM_011519879.2 1117 Silent Mutation XP_011518181.1
XM_017017171.1 1117 Silent Mutation XP_016872660.1
Gene
TRPC2
Gene Name
transient receptor potential cation channel subfamily C member 2, pseudogene
There are no transcripts associated with this gene.

View Full Product Details