Product Details

SNP ID
rs2284462
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:29748806 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGCAAAGAACGATCAACAAATT[A/G]ACACCCTTTTCATATTTTTTGTGAC
Phenotype
MIM: 138245
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIK1 PubMed Links

Gene Details

Gene
GRIK1
Gene Name
glutamate ionotropic receptor kainate type subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000830.4 Intron NP_000821.1
NM_001320616.1 Intron NP_001307545.1
NM_001320618.1 Intron NP_001307547.1
NM_001320621.1 Intron NP_001307550.1
NM_001320630.1 Intron NP_001307559.1
NM_175611.2 Intron NP_783300.1
XM_005260942.1 Intron XP_005260999.1
XM_005260944.3 Intron XP_005261001.1
XM_006723991.1 Intron XP_006724054.1
Gene
GRIK1-AS1
Gene Name
GRIK1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details