Product Details

SNP ID
rs2272166
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197726960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCATCTTGTTTAACATTTGAAAA[C/T]CAAAACATAAACTTTTTGGTTCTCA
Phenotype
MIM: 606165
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BOLL PubMed Links

Gene Details

Gene
BOLL
Gene Name
boule homolog, RNA binding protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284358.1 2694 UTR 3 NP_001271287.1
NM_001284361.1 2694 UTR 3 NP_001271290.1
NM_001284362.1 2694 UTR 3 NP_001271291.1
NM_033030.5 2694 UTR 3 NP_149019.1
NM_197970.2 2694 UTR 3 NP_932074.1
XM_006712715.3 2694 Intron XP_006712778.1
XM_011511692.2 2694 Intron XP_011509994.1
XM_011511693.2 2694 Intron XP_011509995.1
XM_011511694.2 2694 Intron XP_011509996.1
XM_017004773.1 2694 Intron XP_016860262.1

View Full Product Details