Product Details

SNP ID
rs2975129
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:154571024 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCCCCCTCCCCATATCCCCTTC[C/T]TGCTCAATTCCACCTCTCTATACTC
Phenotype
MIM: 118507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHRNB2 PubMed Links

Gene Details

Gene
CHRNB2
Gene Name
cholinergic receptor nicotinic beta 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000748.2 Intron NP_000739.1
XM_017000180.1 Intron XP_016855669.1

View Full Product Details