Product Details

SNP ID
rs2272778
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71241084 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCAGGTTGCAGGTAGAACACATC[A/G]TTGCGAGTCCGGAGGCTTTCAGGAC
Phenotype
MIM: 300061
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZMYM3 PubMed Links

Gene Details

Gene
ZMYM3
Gene Name
zinc finger MYM-type containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171162.1 4133 Silent Mutation AAC,AAT N1303N NP_001164633.1
NM_001171163.1 4133 Intron NP_001164634.1
NM_005096.3 4133 Silent Mutation AAC,AAT N1315N NP_005087.1
NM_201599.2 4133 Silent Mutation AAC,AAT N1315N NP_963893.1
XM_005262309.3 4133 Silent Mutation AAC,AAT N1315N XP_005262366.1
XM_005262310.2 4133 Silent Mutation AAC,AAT N1303N XP_005262367.1
XM_011531062.2 4133 Silent Mutation AAC,AAT N1315N XP_011529364.1

View Full Product Details