Product Details

SNP ID
rs2277929
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:98770321 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTAAGGCCTTCTTGGGAGCGAGTGG[A/C]TGGCTAATGAGAGGTTAATTTTGTT
Phenotype
MIM: 612687
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
RGMB PubMed Links

Gene Details

Gene
RGMB
Gene Name
repulsive guidance molecule family member b
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012761.2 Intron NP_001012779.2
XM_005271966.3 Intron XP_005272023.1
XM_011543345.2 Intron XP_011541647.1
XM_011543346.2 Intron XP_011541648.1
XM_011543347.2 Intron XP_011541649.2
XM_017009390.1 Intron XP_016864879.1
XM_017009391.1 Intron XP_016864880.1
Gene
RGMB-AS1
Gene Name
RGMB antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details